Learn More
Thermo Scientific Chemicals Grade 25 sphére en saphir, Dia. : 1,5 mm
Marque: Thermo Scientific Chemicals 042065.KM
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spécification
Sapphire Sphere | |
1.5 mm | |
Grade 25 | |
25 unités |