Learn More
Thermo Scientific Chemicals Grade 25 sphére en saphir, Dia. : 1,5 mm
Marque:  Thermo Scientific Chemicals 042065.KN
Code produit. 11306019
  • 502.50€ / 100 pièces
Expédition estimée: 16-01-2025
pour voir le stock disponible 
Découvrez plus d'offres spéciales
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Spécification
Spécification
Sapphire Sphere
1.5 mm
Grade 25
100 unités