Learn More
Equip Labo™ Table en grès sans évier
Brand:  Equip Labo™ 8201SECHE
 View more versions of this product
Product Code. 12561487
  • 985.00€ / 1 pièce
Expédition estimée: 24-04-2025
pour voir le stock disponible 
Explore more special offers
Add to basket
This item is not returnable. View return policy
Description
Description
Paillasse sous sorbonne
• Paillasses pour sorbonnes. Equipées d'un rebord en façade conformément à la norme XPX 15-206
• Paillasse pour sorbonne standard et laboratoire
• Paillasse sèche en grès étiré
Specifications
Specifications
Table en grès
1200 x 750 mm
Grès
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.