Learn More
Thermo Scientific™ Sample Storage Starter Packs
Introduce automation into your workflow with Thermo Scientific™ sample storage starter packs
Marque: Thermo Scientific™ HHD-WP1D-2
2275.00 EUR valable jusqu'au 2024-12-31
Utilisez le code promo "23291" pour bénéficier de cette offre.
Code nomenclature Nacres: NB.79
Informations supplémentaires : Poids : 8.00000kg
Description
Introduce automation into your workflow with Thermo Scientific™ sample storage starter packs. Choose from five different starter pack options: BASIC, ADVANCED, PREMIUM, PREMIUM 0.5 mL and PREMIUM 1.mL. All Starter Packs include Matrix™ ScrewTop Tri-coded Tubes with an 8-Channel Screw Cap Decapper. Advanced and Premium Starter Packs also include a barcode reader.
Engineered with the value of your samples in mind:
- Save 40% compared to buying the products individually.
- Choose from five different starter pack options.
- Eliminate the need to reformat non-2D cryotube samples.
- Secure sample tracking with 2D, 1D, and human readable codes.
- Cap/decap a full rack of tubes in less than 1 minute.
- Scan 96 tubes in as little as 1 second with advanced and premium starter packs.
- Help ensure quality with ISO-7 class cleanroom– manufactured tubes with sterility assurance.
Basic Starter Pack Includes:
- 8-Channel Screw Cap Decapper (1 each)
- 0.5 mL Matrix ScrewTop Tri-coded Tubes (1 case)
- 1.0 mL Matrix ScrewTop Tri-coded Tubes (1 case)
Advanced Starter Pack Includes:
- 8-Channel Screw Cap Decapper (1 each)
- VisionMate ST Single-Tube 2D Barcode Reader (1 each)
- 0.5 mL Matrix ScrewTop Tri-coded Tubes (1 case)
- 1.0 mL Matrix ScrewTop Tri-coded Tubes (1 case)
Premium Starter Pack Includes:
- 8-Channel Screw Cap Decapper (1 each)
- VisionMate HSX High Speed Barcode Reader (1 each)
- 0.5 mL Matrix ScrewTop Tri-coded Tubes (1 case)
- 1.0 mL Matrix ScrewTop Tri-coded Tubes (1 case)
Premium 0.5 mL Starter Pack Includes:
- 8-Channel Screw Cap Decapper (1 each)
- VisionMate HSX High Speed Barcode Reader (1 each)
- 0.5 mL Matrix ScrewTop Tri-coded Tubes (5 cases)
Premium 1.0 mL Starter Pack Includes:
- 8-Channel Screw Cap Decapper (1 each)
- VisionMate HSX High Speed Barcode Reader (1 each)
- 1.0 mL Matrix ScrewTop Tri-coded Tubes (5 cases)
Spécification
Basic Starter Pack | |
Tubes and Decapper |