Learn More
Thermo Scientific™ Sample Storage Starter Packs
GREENER_CHOICE
Code produit. 17856148
Click to view available options
Comprend:
Tubes de 0,5 ml, déboucheuse, lecteur
Tubes de 1,0 ml, déboucheuse, lecteur
Tubes et déboucheuse
Tubes, déboucheuse, lecteur HSX
Tubes, déboucheuse, lecteur ST
Conditionnement:
1 kit
Règlementaire : Cet article est soumis à éco-contribution.
Code produit. 17856148
Marque: Thermo Scientific™ HHDHSXWP1D510
Il en reste null
Ajouter au panier
Ajouter au panier
Règlementaire : Cet article est soumis à éco-contribution.
Introduce automation into your workflow with Thermo Scientific™ sample storage starter packs
Introduce automation into your workflow with Thermo Scientific™ sample storage starter packs. Choose from five different starter pack options: BASIC, ADVANCED, PREMIUM, PREMIUM 0.5 mL and PREMIUM 1.mL. All Starter Packs include Matrix™ ScrewTop Tri-coded Tubes with an 8-Channel Screw Cap Decapper. Advanced and Premium Starter Packs also include a barcode reader.
Engineered with the value of your samples in mind:
  • Choose from five different starter pack options.
  • Eliminate the need to reformat non-2D cryotube samples.
  • Secure sample tracking with 2D, 1D, and human readable codes.
  • Cap/decap a full rack of tubes in less than 1 minute.
  • Scan 96 tubes in as little as 1 second with advanced and premium starter packs.
  • Help ensure quality with ISO-7 class cleanroom– manufactured tubes with sterility assurance.
TRUSTED_SUSTAINABILITY
Spécification
Comprend Tubes de 1,0 ml, déboucheuse, lecteur
Type Premium 1.0 mL Starter Pack