missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S100A4 Mouse anti-Human, Mouse, Clone: 2G11B4, Proteintech
Mouse Monoclonal Antibody
Marque: Proteintech 66489-1-IG-20UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
S100 belongs to the family of calcium binding proteins such as calmodulin and troponin C. S100A is composed of an alpha and beta chain whereas S100B is composed of two beta chains. S100 protein is also expressed in the antigen presenting cells such as the Langerhans cells in skin and interdigitating reticulum cells in the paracortex of lymph nodes.Spécification
S100A4 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P07091, P26447 | |
S100A4 | |
S100A4 Fusion Protein ag9019 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG2a |
Immunofluorescence, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
2G11B4 | |
Unconjugated | |
S100A4 | |
18A2, 42A, Calvasculin, CAPL, Fibroblast-specific protein 1, FSP1, FSP-1, Metastasin, MTS1, P9KA, PEL98, Protein Mts1, Protein S100 A4, S100A4 | |
Mouse | |
Protein A | |
RUO | |
20198, 6275 | |
-20°C | |
Liquid |