Learn More
SCAT™ Safety Waste Caps
SafetyWasteCaps SCAT pour une élimination sûre des déchets liquides. Pour une protection maximale et un travail efficace avec les liquides de déchets dangereux. Avec plusieurs options pour connecter différentes tailles de tubes et de capillaires.
Marque: SCAT™ 107121
132.73 EUR valable jusqu'au 2024-09-17
Utilisez le code promo "21690" pour bénéficier de cette offre.
Code nomenclature Nacres: SB.51
Informations supplémentaires : Poids : 0.20400kg
Description
- 360° free turning PTFE body, resistant against aggressive media
- Exhaust filter system against harmful vapors
- Fittings and tube connectors in different sizes
- Flexible system
- Time saving in changing the waste containers while tubes are fixed
- Lab safety, technical protective measures, optimal protection when collecting liquid waste
- Cost efficiency
- Huge range of accessories, optional: funnel use, level control preventing overflowing containers
- Huge portfolio of thread sizes suitable for waste containers from different manufacturers
- Safety disposal of HPLC waste
- High work performance
- Personal protection, avoid skin contact with organic solvents, occupational safety, ergonomics
- Sealed bottles and containers
- Create a comfortable working climate
Spécification
2 x 5 to 11.5 mm ID, Connector for Exhaust Filter | |
S55 | |
White | |
Waste Disposal Container |
HPLC Disposal Systems | |
Safety Waste Cap | |
PTFE Liner |