Learn More
RUVBL2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marque: Invitrogen PA588627
338.68 EUR valable jusqu'au 2025-03-21
Utilisez le code promo "24111" pour bénéficier de cette offre.
Alert:
Pour que la promotion s’applique, le client doit acheter trois fois le même produit au prix catalogue sur la même commande pour avoir une remise de 33.33%. Il n’y a pas de limite d’achat pour un multiple de 3 produits. PCODE: 24111
Description
Immunogen sequence: MATVTATTKV PEIRDVTRIE RIGAHSHIRG LGLDDALEPR QASQGMVGQL AARRAAGVVL EMIREGKIAG RAVLIAGQPG TGKTAIAMGM AQALGPDTPF TAIAGSEIFS LEMSKTEALT QAFRRSIGVR IKEETEIIEG EVVEIQIDRP ATGTGSKVGK LTLKTTEMET IYDLGTKMIE SLTKDKVQAG DVITIDKATG KISKLGRSFT RARDYDAMGS QTKFVQCPDG ELQKRKEVVH TVSLHEIDVI NSRTQGFLAL FSGDTGEIKS EVREQINAKV AEWREEGKAE IIPGVLFIDE VHMLDIESFS FLNRALESDM APVLIMATNR GITRIRGTSY QSPHGIPIDL LDRLLIVSTT PYSEKDTKQI LRIRCEEEDV EMSEDAYTVL TRIGLETSLR YAIQLITAAS LVCRKRKGTE VQVDDIKRVY SLFLDESRST QYMKEYQDAF LFNELKGETM DTS; Positive Samples: BT-474, K-562, SW480, MCF7 ; Cellular Location: Cytoplasm, Membrane, Nucleus, Nucleus matrix, nucleoplasm
Possesses single-stranded DNA-stimulated ATPase and ATP-dependent DNA helicase (5' to 3') activity. Component of the NuA4 histone acetyltransferase complex which is involved in transcriptional activation of select genes principally by acetylation of nucleosomal histones H4 and H2A. This modification may both alter nucleosome - DNA interactions and promote interaction of the modified histones with other proteins which positively regulate transcription. This complex may be required for the activation of transcriptional programs associated with oncogene and proto-oncogene mediated growth induction, tumor suppressor mediated growth arrest and replicative senescence, apoptosis, and DNA repair. The NuA4 complex ATPase and helicase activities seem to be, at least in part, contributed by the association of RUVBL1 and RUVBL2 with EP400. NuA4 may also play a direct role in DNA repair when recruited to sites of DNA damage. RUVBL2 plays an essential role in oncogenic transformation by MYC and also modulates transcriptional activation by the LEF1/TCF1-CTNNB1 complex. May also inhibit the transcriptional activity of ATF2.
Spécification
RUVBL2 | |
Polyclonal | |
Unconjugated | |
Ruvbl2 | |
48 kDa TATA box-binding protein-interacting protein, 48 kDa TBP-interacting protein, 51 kDa erythrocyte cytosolic protein, CGI-46, ECP51, ECP-51, erythrocyte cytosolic protein, 51-KD, INO80 complex subunit J, INO80J, liebeskummer, lik, mp47, p47, p47 protein, Repressing pontin 52, reptin, Reptin 52, reptin52 protein, RuvB (E coli homolog)-like 2, RuvB like AAA ATPase 2, RuvB like AAA ATPase 2 S homeolog, ruvbl2, RUVBL2 protein, ruvbl2.S, ruvB-like 2, RuvB-like AAA ATPase 2, RuvB-like DNA helicase reptin, RuvB-like protein 2, RVB2, TAP54-beta, TIH2, TIP48, TIP49B, TIP60-associated protein 54-beta, wu:fi25f01, XELAEV_180378471mg, xReptin, zReptin | |
Rabbit | |
Affinity Chromatography | |
RUO | |
10856, 20174, 292907 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot, Immunohistochemistry, Western Blot | |
0.25 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide, pH 7.3 | |
Q9WTM5, Q9Y230 | |
Ruvbl2 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-463 of human Reptin/Reptin/RUVBL2 (NP_0066571) | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.