Learn More
Invitrogen™ RSV Polyclonal Antibody, HRP
Goat Polyclonal Antibody
Marque: Invitrogen™ PA173018
Description
The PA1-73018 antibody reacts with Respiratory Syncytial virus. PA1-73018 has been successfully used in ELISA applications. The PA1-73018 antibody was raised against Human RSV isolate (confirmed). Store at 4°C for up to 6 months. For extended storage, store at -20°C, avoiding freeze/thaw cycles.
Human respiratory syncytial virus (RSV) is a member of the Paramyxoviridae family, subfamily Pneumovirinae, that causes common respiratory tract infections such as measles and mumps. Its genome consists of single stranded, negative sense RNA that encodes three envelope glycoproteins, a small hydrophobic (SH) protein of unknown function, a major glycoprotein (G) known as the attachment protein, and a fusion (F) protein. Its name comes from the fact that F proteins on the surface of the virus cause the membranes on nearby cells to merge, forming syncytia. While RSV typically produces mild symptoms in adults, it is one of the leading causes of lower respiratory tract infections and hospital visits during infancy and childhood.
Spécification
| RSV | |
| Polyclonal | |
| HRP | |
| human respiratory syncytial virus; RSV | |
| Human RSV isolate (confirmed). | |
| 1 mL | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| PBS with 10mg/mL BSA and 0.002% thimerosal | |
| Goat | |
| IgG fraction | |
| RUO | |
| Virus | |
| Antibody | |
| IgG |
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.