Learn More
RPS6KA1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque:  Proteintech 16463-1-AP-20UL
Code produit. 16846094
Description
Description
This gene encodes a zinc finger protein of unknown function. It bears similarity to a zinc finger protein which acts as a transcriptional activator. This gene lies in a region of the X chromosome which has been associated with mental retardation.
Spécification
Spécification
RPS6KA1
Polyclonal
Unconjugated
RPS6KA1
HU 1, MAPKAP kinase 1a, MAPKAPK 1a, MAPKAPK1A, p90 RSK 1, p90RSK1, p90S6K, Ribosomal S6 kinase 1, RPS6KA1, RSK, RSK 1, RSK1, S6K alpha 1
Rabbit
Antigen Affinity Chromatography
RUO
20111, 6195, 81771
-20°C
Liquid
Western Blot
0.13 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P18653, Q15418, Q63531
RPS6KA1
RPS6KA1 Fusion Protein Ag9537
20 μL
Primary
Human, Rat, Mouse
Antibody
IgG
Nom du produit
RPS6KA1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated