missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPS25 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 23599-1-AP-150UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
23599-1-AP is a rabbit polyclonal antibody raised against full-length RPS25, and is recommended for ELISA, Western blotting and IP assays.
Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 40S subunit. The protein belongs to the S25E family of ribosomal proteins. It is located in the cytoplasm.Spécification
RPS25 | |
Polyclonal | |
Unconjugated | |
RPS25 | |
40S ribosomal protein S25, ribosomal protein S25, RPS25 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
6230 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.45 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P62851 | |
RPS25 | |
RPS25 Fusion Protein Ag7890 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |