missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ro 51, Tocris Bioscience™
Click to view available options
Quantité:
10 mg
50 mg
Description
Potent, dual P2X3 and P2X2/3 antagonist (IC50 values are 2 and 5 nM for rP2X3 and hP2X2/3 respectively). Selective for P2X3 and P2X2/3 over other P2X receptors (IC50 values are > 10 μM for P2X1, P2X2, P2X4, P2X5, and P2X7). Cell permeable.
Spécification
Spécification
Nom chimique ou matériau | Ro 51 |
Quantité | 10 mg |
Poids de la formule | 474.29 |
Pourcentage de pureté | >98% |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'