missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RNase T2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 13753-1-AP-150UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
RNASET2 is a novel member of the Rh/T2/S-glycoprotein class of extracellular ribonucleases. It is a single copy gene that maps to 6q27, a region associated with human maligcies and chromosomal rearrangement, and has been suggested to function as a tumor suppressor protein. Its expression is suppressed in Human T-cell Leukemia Virus type 1 (HTLV-1) infected cells following the binding of the HTLV-1 Tax protein to the RNASET2 promoter. As Adult T-cell leukemia (ATL) is one of the primary diseases caused by HTLV-1 infection, a reduction in the level of RNASET2 by Tax may play a role in ATL development.Spécification
RNase T2 | |
Polyclonal | |
Unconjugated | |
RNASET2 | |
Ribonuclease 6, Ribonuclease T2, RNASE6PL, RNASET2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
100037283, 8635 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
C0HKG5, O00584 | |
RNASET2, Rnaset2a | |
RNase T2 Fusion Protein Ag4695 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |