Learn More
Corning™ rLaminin-521 (Human)
Tous les produits Corning ProduitsDescription
Corning extracellular matrices (ECMs) enable researchers to mimic in vivo environments for 2D and 3D cell culture applications.
Corning rLaminin-521 is the next generation pluripotent stem cell (PSC) surface that enables researchers to streamline culture workflow, lower contamination risk, and scale-up efficiently. Corning has partnered with BioLamina for the supply of recombinant human Laminin 521 (rLaminin-521), a biocompatible, full length Laminin that enables feeder-free, clump-free, single-cell passaging of PSCs without ROCK-inhibitor.
Spécification
Spécification
| Nom | rLaminin-521, Human |
| Quantité | 5 mg |
| Source | Human |
| Recombinant | Recombinant |