Learn More
Labconco™ ReVo MSC IV Bar
Code produit. 17835602 Tous les produits Labconco Corporation Produits
Click to view available options
Dimensions (L x l x H) extérieures:
900 x 20 x 20 mm
1 200 x 20 x 20 mm
1500 x 20 x 20 mm
1800 x 20 x 20 mm
Conditionnement:
1 unité
Code produit. 17835602
Marque: Labconco™ R3858601
Bar supports intravenous solution bottles and bags. Kit includes IV bar and mounting hardware.
  • IV Bar and mounting hardware compatible with various biosafety cabinets, including Purifier Axiom Biosafety Cabinets, Purifier Cell Logic and Cell Logic+ Biosafety Cabinets, Purifier Logic and Logic+ Biosafety Cabinets, Purifier Logic and PuriCare Procedure Stations and ReVo Microbiological Safety Cabinets.
Spécification
Type de produit IV Bag Holder Kit
Dimensions (L x l x H) extérieures 1 200 x 20 x 20 mm
Poids à l’expédition (métrique) 2.0 kg