Learn More
Labconco™ ReVo Canopy Connection Kit
ReVo Canopy Connection Kit is ideal for ducting ReVo Class II, Type A2 Microbiological Safety Cabinets.
Marque:  Labconco™ R3889212
Code produit. 17875592
  • 1370.00€ / 1 kit
Découvrez plus d'offres spéciales
Description
Description
  • Canopy Connection Kit with side exhaust for ducting ReVo Class II, Type A2 Microbiological Safety Cabinets.
Spécification
Spécification
Canopy Connection Kit
465 x 680 x 265 mm
1500/1800 mm Cabinet
5.0 kg