Learn More
REG1 alpha (REG1A) Mouse anti-Human, Clone: OTI11E1, TrueMAB™, Origene
Mouse Monoclonal Antibody
Marque: Origene Technologies TA813908S
Description
REG1 alpha (REG1A) Monoclonal antibody specifically detects REG1 alpha (REG1A) in Human samples. It is validated for ELISASpécification
| REG1 alpha (REG1A) | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P05451 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 5967 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA | |
| OTI11E1 | |
| Unconjugated | |
| REG1A | |
| ICRF; islet cells regeneration factor; Islet of Langerhans regenerating protein; LITHOST; Lithostathine; lithostathine 1 alpha; lithostathine-1-alpha; MGC12447; P19; pancreatic stone protein; pancreatic stone protein, secretory; pancreatic thread protein; protein-X; PSP; PSPS; PSPS1; PTP; RATLITHOST; RATRGPI; Reg; Reg1; REG1A; REG-1-alpha; regenerating family member 1 alpha; regenerating islet-derived 1 alpha; regenerating islet-derived mouse homolog 1; regenerating islet-derived protein 1-alpha; Regenerating protein I alpha; regeneration protein, lithostatin, pancreatic stone protein; Rgp1; RGPI | |
| Human recombinant protein fragment corresponding to amino acids 23-166 of human PSP (NP_002900) produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
Le programme Encompass de Fisher Scientific propose des articles qui ne font pas partie de notre portefeuille de distribution. Ces produits n'ont généralement pas de photos ou de descriptions détaillées. Cependant, nous nous engageons à améliorer votre expérience d'achat. Veuillez utiliser le formulaire ci-dessous pour nous faire part de vos commentaires sur le contenu de ce produit.