missing translation for 'onlineSavingsMsg'
Learn More

REEP6 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marque:  Invitrogen PA561635

Code produit. 15779441

  • 478.00€ / 100µL

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

Découvrez plus d'offres spéciales
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Description

Description

Immunogen sequence: MDGLRQRVEH FLEQRNLVTE VLGALEAKTG VEKRYLAAGA VT Highest antigen sequence identity to the following orthologs: Mouse - 79%, Rat - 79%.

REEP6 may enhance the cell surface expression of odorant receptors.
TRUSTED_SUSTAINABILITY
Spécification

Spécification

REEP6
Polyclonal
Unconjugated
REEP6
0610011M24Rik; C19orf32; deleted in polyposis 1-like 1; DP1L1; Polyposis locus protein 1-like 1; receptor accessory protein 6; receptor expression enhancing protein 6; receptor expression-enhancing protein 6; REEP6; TB2 protein-like 1; Tb2l1; Yip2f
Rabbit
Antigen affinity chromatography
RUO
92840
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunocytochemistry, Immunohistochemistry, Western Blot
0.4 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q96HR9
REEP6
Recombinant protein corresponding to Human REEP6
100 μL
Primary
Human
Antibody
IgG
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions

Research Use Only