Learn More
Gilac™ Rectangular HACCP Container with Lid
Click to view available options
Couleur:
Blanc
Jaune
Rouge
Vert
Capacité (métrique):
12 l
15 l
35 l
55 l
Conditionnement:
1 pièce
Description
- HACCP rectangular food container.
- For large storage volumes, lid can support the weight of heavy loads.
- Multipurpose container.
- Rewritable identification cartridge for inventory tracking.
- No gaps nor overlaps.
- Machine washable.
- Recessed handles for better handling.
- Stackable without lid.
- Compatible with all GILAC 4 and 6 wheel dollies.
Spécification
Spécification
| Type | Storage Tote |
| Matériau | HDPE |
| Couleur | Rouge |
| Forme | Rectangulaires |
| Capacité (impériale) | 4 gal. |
| Capacité (métrique) | 15 l |