Learn More
R&D Systems™ Reconstitution Buffer 4 (BSA/HCl)
missing translation for 'orderingAttributeHoverText'
Quantité:
2 ml
missing translation for 'unitSize'
1 pièce
Description
This product is intended as a companion product for reconstitution of selected lyophilized antibody or protein to maximize recovery and activity. Refer to the specific antibody or protein product data sheet for the recommended reconstitution concentrations.
Specifications
Specifications
| Quantité | 2 ml |