missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rabbit IL17 / IL17A Protein
A DNA sequence encoding the Oryctolagus cuniculus (Rabbit) IL17 / IL17A, was expressed in the hosts and tags indicated.
Marque: enQuireBio™ QP6212-ye-500ug
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Spécification
XP_002714544.1 | |
IL17 / IL17A Protein | |
Research Use Only | |
IL17A | |
Recombinant Protein | |
Yeast | |
VKNGIAMPRN PGCPNAEDKN FPQNVKVSLN ILNKSVNSRR PSDYYNRSTS PWTLHRNEDR ERYPSVIWEA KCRHLGCVNA EGNEDHHMNS VPIQQEILVL RRESQHCPHS FRLEKMLVAV GCTCVTPIIH HMA | |
Greater than 90% as determined by SDS-PAGE. |
100339322 | |
500 μg | |
Not determined. | |
Full Length | |
Rabbit | |
His | |
Tris-based buffer, 50% glycerol |