missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Pig ETFDH Protein
A DNA sequence encoding the Sus scrofa (Pig) ETFDH, was expressed in the hosts and tags indicated.
Marque: enQuireBio™ QP6003-ye-200ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
ETFDH Protein | |
Research Use Only | |
ETFDH | |
Recombinant Protein | |
Yeast | |
SSTCKVPRIT THYTIYPRDQ DKRWEGVNME RFAEEADVVI VGAGPAGLSA ATRLKQLAAQ HEKDLRVCLV EKAAHIGAHT LSGACLDPRA FEELFPDWKE KGAPLNTPVT EDRFGILTEK YRIPVPILPG LPMNNHGNYV VRLGHLVSWM GEQAEALGVE VYPGYAAAEI LFHEDGSVKG IATNDVGIQK DGAPKTTFER GLELHAKVTI FAEGCHGHLA KQLYKKFDLR ANCEPQTYGI GLKELWVIDE KKWKPGRVDH TVGWPLDRHT YGGSFLYHLN EGEPLLALGF VVGLDYQNPY LSPFREFQRW KHHPSIKPTL EGGKRIAYGA RALNEGGFQS IPKLTFPGGL LIGCSPGFMN VPKIKGTHTA MKSGTLAAES IFNQLTSENL QSKTIGLHVT EYEDNLKNSW VWKELYSVRN IRPSCHGILG VYGGMIYTGI FYWIFRGMEP WTLKHKGSDS DQLKPAKDCT PIEYPKPDGQ ISFDLLSSVA LSGTNHEHDQ PAHLTLKDDS VPVNRNLSIY DGPEQRFCPA GVYEFVPLEQ GDGFRLQINA QNCVHCKTCD IKDPSQNINW VVPEGGGGPA YNGM | |
Greater than 85.3% as determined by SDS-PAGE. |
200 μg | |
Not determined. | |
Full Length | |
Pig | |
His | |
Tris-based buffer, 50% glycerol |