missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant other Placental Lactogen Caprine Protein

A cDNA sequence encoding the Placental Lactogen Caprine was constructed and used to recombinantly synthesize the protein.

Marque:  enQuireBio™ QP10828-200ug

 Afficher plus de versions de ce produit

Code produit. 15995288

  • 203.00€ / 200µg

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

Découvrez plus d'offres spéciales
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Description

Description

Spécification

Spécification

Placental Lactogen Caprine Protein
Research Use Only
Placental Lactogen Caprine is biologically active as evidenced by inducing proliferation of Nb2 cells.
Other
Untagged
Greater than 98.0% as determined by:(a) Analysis by SEC-HPLC. (b) Analysis by SDS-PAGE.
200 μg
< 1.0 EU per ug protein as determined by the LAL method.
Recombinant Protein
E. coli
The protein was lyophilized from a concentrated (1 mg/ml) solution with 0.02-0.03% NaHCO3.
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions