missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other Placental Lactogen Caprine Protein
A cDNA sequence encoding the Placental Lactogen Caprine was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP10828-50ug
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Spécification
Placental Lactogen Caprine Protein | |
Research Use Only | |
Placental Lactogen Caprine is biologically active as evidenced by inducing proliferation of Nb2 cells. | |
Other | |
Untagged | |
Greater than 98.0% as determined by:(a) Analysis by SEC-HPLC. (b) Analysis by SDS-PAGE. |
50 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
The protein was lyophilized from a concentrated (1 mg/ml) solution with 0.02-0.03% NaHCO3. |