missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse IL34 Protein

A cDNA sequence encoding the IL34 was constructed and used to recombinantly synthesize the protein.

Marque:  enQuireBio™ QP12422-1mg

Code nomenclature Nacres: NA.28

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15980369

  • 7540.00€ / 1mg
Expédition estimée: 05-09-2024
pour voir le stock disponible 

Description

Description

Spécification

Spécification

76527
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MNENLEIWTL TQDKECDLTG YLRGKLQYKN RLQYMKHYFP INYRIAVPYE GVLRVANITR LQKAHVSERE LRYLWVLVSL NATESVMDVL LEGHPSWKYL QEVQTLLENV QRSLMDVEIG PHVEAVLSLL STPGLSLKLV RPKALLDNCF RVMELLYCSC CKQSPILKWQ DCELPRLHPH SPGSLMQCTA TNVYPLSRQT PTSLPGSPSS SHGSLP
Greater than 85% as determined by SDS-PAGE.
IL34 Protein
Research Use Only
Il34
Mouse
His
IL34 protein solution (0.5 mg/ml) in 20mM Tris-HCl buffer (pH 8.0) and 10% Glycerol.
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions