missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human UPK2 Protein
A cDNA sequence encoding the UPK2 was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP13897-5ug
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Spécification
UPK2 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSDFNISSL SGLLSPALTE SLLVALPPCH LTGGNATLMV RRANDSKVVT SSFVVPPCRG RRELVSVVDS GAGFTVTRLS AYQVTNLVPG TKFYISYLVK KGTATESSRE IPMSTLPRRN MESIGLGMAR TGG | |
Greater than 95.0% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
UPK2 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 2M UREA and 10% glycerol. |