missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Sterol regulatory element-binding protein 1
A DNA sequence encoding the Homo sapiens(Human) Sterol regulatory element-binding protein 1, was expressed in the hosts and tags indicated.
469.00€ - 1630.00€
Spécification
Numéro d’adhésion | NP_001005291.1 |
---|---|
Identification génétique (Entrez) | 6720 |
Nom | Sterol regulatory element-binding protein 1 |
État réglementaire | Research Use Only |
Concentration en endotoxines | Not determined. |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
16019141
|
enQuireBio™
QP9983-iv-10ug |
10 μg | N/A | ||||||
16029141
|
enQuireBio™
QP9983-iv-50ug |
50 μg | N/A | ||||||
16009141
|
enQuireBio™
QP9983-iv-100ug |
100 μg |
1630.00€
100µg |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | |||||
Specifications
NP_001005291.1 | |
Sterol regulatory element-binding protein 1 | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
6720 | |
Research Use Only | |
SREBF1 | |
Recombinant Protein | |
E. coli | |
MDEPPFSEAA LEQALGEPCD LDAALLTDIE DMLQLINNQD SDFPGLFDPP YAGSGAGGTD PASPDTSSPG SLSPPPATLS SSLEAFLSGP QAAPSPLSPP QPAPTPLKMY PSMPAFSPGP GIKEESVPLS ILQTPTPQPL PGALLPQSFP APAPPQFSST PVLGYPSPPG GFSTGSPPGN TQQPLPGLPL ASPPGVPPVS LHTQVQSVVP QQLLTVTAAP TAAPVTTTVT SQIQQVPVLL QPHFIKADSL LLTAMKTDGA TVKAAGLSPL VSGTTVQTGP LPTLVSGGTI LATVPLVVDA EKLPINRLAA GSKAPASAQS RGEKRTAHNA IEKRYRSSIN DKIIELKDLV VGTEAKLNKS AVLRKAIDYI RFLQHSNQKL KQENLSLRTA VHKSKSLKDL VSACGSGGNT DVLMEGVKTE VEDTLTPPPS DAGSPFQSSP LSLGSRGSGS GGSGSDSEPD SPVFEDSKAK PEQRPSLHSR GMLDRSRLAL | |
Greater than 85% as determined by SDS-PAGE. |