missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SP-10 / ACRV1 Protein
A DNA sequence encoding the Homo sapiens (Human) SP-10 / ACRV1, was expressed in the hosts and tags indicated.
Marque: enQuireBio™ QP5609-ec-1mg
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Spécification
NP_001603.1 | |
SP-10 / ACRV1 Protein | |
Research Use Only | |
ACRV1 | |
Recombinant Protein | |
E. coli | |
QPNELSGSID HQTSVQQLPG EFFSLENPSD AEALYETSSG LNTLSEHGSS EHGSSKHTVA EHTSGEHAES EHASGEPAAT EHAEGEHTVG EQPSGEQPSG EHLSGEQPLS ELESGEQPSD EQPSGEHGSG EQPSGEQASG EQPSGEHASG EQASGAPISS TSTGTILNCY TCAYMNDQGK CLRGEGTCIT QNSQQCMLKK IFEGGKLQFM VQGCENMCPS MNLFSHGTRM QIICCRNQSF CNKI | |
Greater than 90% as determined by SDS-PAGE. |
56 | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |