missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human RASL12 Protein
A cDNA sequence encoding the RASL12 was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP13272-1mg
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Spécification
RASL12 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMSSVFGK PRAGSGPQSA PLEVNLAILG RRGAGKSALT VKFLTKRFIS EYDPNLEDTY SSEETVDHQP VHLRVMDTAD LDTPRNCERY LNWAHAFLVV YSVDSRQSFD SSSSYLELLA LHAKETQRSI PALLLGNKLD MAQYRQVTKA EGVALAGRFG CLFFEVSACL DFEHVQHVFH EAVREARREL EKSPLTRPLF ISEERALPHQ APLTARHGLA SCTFNTLSTI NLKEMPTVAQ AKLVTVKSSR AQSKRKAPTL TLLKGFKIF | |
Greater than 85.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
RASL12 protein solution (0.5 mg/ml) containing 20mM Tirs-HCl (pH 8.0) and 10% glycerol. |