missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PLAC8 Protein
A cDNA sequence encoding the PLAC8 was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP13075-25ug
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Spécification
PLAC8 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMQAQAPV VVVTQPGVGP GPAPQNSNWQ TGMCDCFSDC GVCLCGTFCF PCLGCQVAAD MNECCLCGTS VAMRTLYRTR YGIPGSICDD YMATLCCPHC TLCQIKRDIN RRRAMRTF | |
Greater than 85.0% as determined by SDS-PAGE. |
25 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
PLAC8 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0),0.4M urea and 10% glycerol. |