missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human NTAL Protein
A cDNA sequence encoding the NTAL was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP12891-25ug
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Spécification
NTAL Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MRCSRPGAKR SEKIYQQRSL REDQQSFTGS RTYSLVGQAW PGPLADMAPT RKDKLLQFYP SLEDPASSRY QNFSKGSRHG SEEAYIDPIA MEYYNWGRFS KPPEDDDANS YENVLICKQK TTETGAQQEG IGGLCRGDLS LSLALKTGPT SGLCPSASPE EDEESEDYQN SASIHQWRES RKVMGQLQRE ASPGPVGSPD EEDGEPDYVN GEVAATEA | |
Greater than 80% as determined by SDS-PAGE. |
25 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
NTAL Human solution containing 20mM Trsi HCL pH 8.0, and 10% glycerol. |