missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human LSM2 Protein
A cDNA sequence encoding the LSM2 was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP12591-5ug
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Spécification
LSM2 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMLFYSF FKSLVGKDVV VELKNDLSIC GTLHSVDQYL NIKLTDISVT DPEKYPHMLS VKNCFIRGSV VRYVQLPADE VDTQLLQDAA RKEALQQKQ | |
Greater than 90% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The LSM2 solution (0.5 mg/1ml) contains 20mM Tris-HCl buffer (pH 8.0), 200mM Nacl, 5mM DTT and 40% glycerol. |