missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human KCNMB3 Protein
A cDNA sequence encoding the KCNMB3 was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP12479-20ug
Code nomenclature Nacres: NA.28
Informations supplémentaires : Poids : 0.01000kg
Spécification
KCNMB3 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSKPFMLSI QREESTCTAI HTDIMDDWLD CAFTCGVHCH GQGKYPCLQV FVNLSHPGQK ALLHYNEEAV QINPKCFYTP KCHQDRNDLL NSALDIKEFF DHKNGTPFSC FYSPASQSED VILIKKYDQ | |
Greater than 85.0% as determined by SDS-PAGE. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
KCNMB3 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.4M UREA and 10% glycerol. |