missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human HMBS Protein
A cDNA sequence encoding the HMBS was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP12279-5ug
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Spécification
HMBS Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMSGNGN AAATAEENSP KMRVIRVGTR KSQLARIQTD SVVATLKASY PGLQFEIIAM STTGDKILDT ALSKIGEKSL FTKELEHALE KNEVDLVVHS LKDLPTVLPP GFTIGAICKR ENPHDAVVFH PKFVGKTLET LPEKSVVGTS SLRRAAQLQR KFPHLEFRSI RGNLNTRLRK LDEQQEFSAI ILATAGLQRM GWHNRVGQIL HPEECMYAVG QGALGVEVRA KDQDILDLVG VLHDPETLLR CIAERAFLRH LEGGCSVPVA VHTAMKDGQL YLTGGVWSLD GSDSIQETMQ ATIHVPAQHE DGPEDDPQLV GITARNIPRG PQLAAQNLGI SLANLLLSKG AKNILDVARQ LNDAH | |
Greater than 95.0% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The HMBS solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 1mM DTT, 10% glycerol and 0.1M NaCl. |