missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human HLA DMB GST (N-Term) Protein
Recombinant Protein
Marque: Novus Biologicals™ H00003109-Q01-25ug
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
This protein was produced in an in vitro wheat germ expression system that should preserve correct conformational folding that is necessary for biological function. While it is possible that this protein could display some level of activity, the functionality of this protein has not been explicitly measured or validated.
Spécification
Western Blot, ELISA, Protein Array, Immunoaffinity Purification | |
3109 | |
25 μg | |
class II histocompatibility antigen, M beta chain, D6S221E, DMB, major histocompatibility complex, class II, DM beta, MHC class II antigen DMB, MHC class II antigen HLA-DM beta chain, MHC class II HLA-DMB, Really interesting new gene 7 protein, RING7HLA class II histocompatibility antigen, DM beta chain | |
Asthma, Diabetes Research, Immunology | |
HLA DMB |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer. | |
37.62 kDa | |
Store at -80°C. Avoid freeze-thaw cycles. | |
HLA-DMB | |
>80% by SDS-PAGE and Coomassie blue staining |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Novus Biologicals™ Recombinant Human HLA DMB GST (N-Term) Protein > 25 μg
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu