missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human FTSJ1 Protein
A DNA sequence encoding the Homo sapiens (Human) FTSJ1, was expressed in the hosts and tags indicated.
Marque: enQuireBio™ QP6058-ec-500ug
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Spécification
NP_001269086.1 | |
FTSJ1 Protein | |
Research Use Only | |
FTSJ1 | |
Recombinant Protein | |
E. coli | |
MGRTSKDKRD VYYRLAKENG WRARSAFKLL QLDKEFQLFQ GVTRAVDLCA APGSWSQVLS QKIGGQGSGH VVAVDLQAMA PLPGVVQIQG DITQLSTAKE IIQHFKGCPA DLVVCDGAPD VTGLHDVDEY MQAQLLLAAL NIATHVLKPG GCFVAKIFRG RDVTLLYSQL QVFFSSVLCA KPRSSRNSSI EAFAVCQGYD PPEGFIPDLS KPLLDHSYDP DFNQLDGPTR IIVPFVTCGD LSSYDSDRSY PLDLEGGSEY KYTPPTQPPI SPPYQEACTL KRKGQLAKEI RPQDCPISRV DTFPQPLAAP QCHTLLAPEM EDNEMSCSP | |
Greater than 90% as determined by SDS-PAGE. |
24140 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |