missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human Enolase 2/Neuron-specific Enolase Protein
Tous les produits Bio Techne Produits
Click to view available options
Quantité:
1 mg
100 μg
5 μg
500 μg
Conditionnement:
100µg
1mg
500µg
5µg
Spécification
Spécification
| Formule | Lyophilized from a 0.2 μm filtered concentrated solution in PBS, pH 7.4. |
| Identification génétique (Entrez) | 2026 |
| Poids moléculaire | MolecularWeight-theroretical: 47.1 kDa |
| Quantité | 5 μg |
| Conditions de stockage | Store at -20°C to -70°C as supplied. After reconstitution, store at 2 to 8 C for 1 month and (at -20°C to -70°C for long term storage. Avoid repeated freeze/thaw cycles.) |
| Concentration en endotoxines | Less than 0.1 EU/μg of Enolase 2/Neuron-specific Enolase as determined by LAL method. |
| Alias de gène | 2-phospho-D-glycerate hydrolyase, 2-phospho-D-glycerate hydro-lyase, EC 4.2.1.11, Enolase 2, enolase 2 (gamma, neuronal), gamma-enolase, Neural enolase, neuron specific gamma enolase, neurone-specific enolase, Neuron-specific enolase, NSE |
| Symbole de gène(s) | ENO2 |
| Reconstitution | Recommended to centrifuge prior to opening. Reconstitute in sterile distilled water or aqueous buffer containing 0.1% BSA to a concentration of 0.1-1.0 mg/mL Apportion stock solutions into working aliquots and store (at <-20°C.) |
| Catégorie de recherche | Cancer, Cellular Markers, Neuronal Cell Markers, Neuroscience |
| Afficher plus |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu