missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human DMC1 GST (N-Term) Protein
Recombinant Protein
611.00€ - 812.00€
Specifications
À utiliser avec (application) | Western Blot, ELISA, Protein Array, Immunoaffinity Purification |
---|---|
Formule | 50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer. |
Identification génétique (Entrez) | 11144 |
Poids moléculaire | 37.07 kDa |
Conditions de stockage | Store at -80°C. Avoid freeze-thaw cycles. |
Product Code | Brand | Quantité | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantité | Price | Quantity & Availability | |||||
18317969
|
Novus Biologicals™
H00011144-Q01-10ug |
10 μg |
611.00€
10µg |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | |||||
18373109
|
Novus Biologicals™
H00011144-Q01-25ug |
25 μg |
812.00€
25µg |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | |||||
Description
This protein was produced in an in vitro wheat germ expression system that should preserve correct conformational folding that is necessary for biological function. While it is possible that this protein could display some level of activity, the functionality of this protein has not been explicitly measured or validated.Specifications
Western Blot, ELISA, Protein Array, Immunoaffinity Purification | |
11144 | |
Store at -80°C. Avoid freeze-thaw cycles. | |
DMC1 | |
>80% by SDS-PAGE and Coomassie blue staining |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer. | |
37.07 kDa | |
disrupted meiotic cDNA1, yeast, homolog of, DMC1 dosage suppressor of mck1 homolog, meiosis-specific homologousrecombination (yeast), DMC1 homologue, HsLim15, LIM15yeast homolog) meiosis-specific homologousrecombination, meiotic recombination protein DMC1/LIM15 homolog, MGC150472, MGC150473 | |
Apoptosis, Cancer, Cell Cycle and Replication, DNA Repair, Homologous Recombination, Tumor Suppressors | |
DMC1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Novus Biologicals™ Recombinant Human DMC1 GST (N-Term) Protein