missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human BMP13 Protein
A cDNA sequence encoding the BMP13 was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP10537-1mg
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Spécification
392255 | |
1 mg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
The ED50 as determined by inducing alkaline phosphatase production of murine ATDC5 cells is less than 2.0microg/ml, corresponding to a specific activity of > 500IU/mg. | |
Human | |
Untagged | |
BMP-13 protein was lyophilized from a 0.2microm filtered concentrated solution in 30% Acetonitrile and 0.1% TFA. |
BMP13 Protein | |
Research Use Only | |
GDF6 | |
Recombinant Protein | |
E. coli | |
TAFASRHGKR HGKKSRLRCS KKPLHVNFKE LGWDDWIIAP LEYEAYHCEG VCDFPLRSHL EPTNHAIIQT LMNSMDPGST PPSCCVPTKL TPISILYIDA GNNVVYKQYE DMVVESCGCR | |
Greater than 95.0% as determined by: (a) Analysis by HPLC. (b) Analysis by SDS-PAGE. |