missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human AZGP1 Protein
A cDNA sequence encoding the AZGP1 was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP10527-2ug
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Spécification
563 | |
2 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
Human (HEK293) | |
ASWSHPQFEK GSQENQDGRY SLTYIYTGLS KHVEDVPAFQ ALGSLNDLQF FRYNSKDRKS QPMGLWRQVEGMEDWKQDSQ LQKAREDIFM ETLKDIVEYY NDSNGSHVLQ GRFGCEIENN RSSGAFWKYY YDGKDYIEFNKEIPAWVPFD PAAQITKQKW EAEPVYVQRA KAYLEEECPA TLRKYLKYSK NILDRQDPPS VVVTSHQAPG EKKKLKCLAYDFYPGKIDVH WTRAGEVQEP ELRGDVLHNG NGTYQSWVVV AVPPQDTAPY SCHVQHSSLA QPLVVPWEAS | |
Greater than 90.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |
AZGP1 Protein | |
Research Use Only | |
AZGP1 | |
Human | |
Untagged | |
Filtered (0.4 microm) and lyophilized in 0.5 mg/ml in 0.1M Tris-HCl pH 8.0 and 150mM NaCl. |