missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Chimpanzee Glypican-3 Protein
A DNA sequence encoding the Pan troglodytes (Chimpanzee) Glypican-3, was expressed in the hosts and tags indicated.
Marque: enQuireBio™ QP10081-ma-100ug
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Spécification
NP_001129209.1 | |
Glypican-3 Protein | |
Research Use Only | |
GPC3 | |
Recombinant Protein | |
Mammal | |
QPPPPPPDAT CHQVRSFFQR LQPGLKWVPE TPVPGSDLQV CLPKGPTCCS RKMEEKYQLT ARLNMEQLLQ SASKELKFLI IQNAAVFQEA FEIVVRHAKN YTNAMFKNNY PSLTPQAFEF VGEFFTDVSL YILGSDINVD DMVNELFDSL FPVIYTQLMN PGLPDSALDI NECLRGARRD LKVFGNFPKL IMTQVSKSLQ VTRIFLQALN LGIEVINTTD HLKFSKDCGR MLTRMWYCSY CQGLMMVKPC GGYCNVVMQG CMAGVVEIDK YWREYILSLE ELVNGMYRIY DMENVLLGLF STIHDSIQYV QKNAGKLTTT IGKLCAHSQQ RQYRSAYYPE DLFIDKKVLK VARVEHEETL SSRRRELIQK LKSFISFYSA LPGYICSHSP VAENDTLCWN GQELVERYSQ KAARNGMKNQ FNLHELKMKG PEPVVSQIID KLKHINQLLR TMSVPKGRVL DKNLDEEGFE SGDCGDDEDE CIGGSGDGMM KVKNQLRFLA ELAYDLDVDD APGSNQQATP KDNEISTFHN LGNVHS | |
Greater than 90% as determined by SDS-PAGE. |
465866 | |
100 μg | |
Not determined. | |
Full Length | |
Chimpanzee | |
His | |
Tris-based buffer, 50% glycerol |