Learn More
Elma™ Reaction Tube Holders
Ensure that reaction tubes can be securely positioned and arranged in the Elma Basket.
Marque: Elma™ 1112152
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
- Optimum positioning of the items inside the bath is also a prerequisite for achieving the best possible cleaning results
- Allows items to be individually positioned
- Lets you clearly structure your cleaning process
- Prevents scratches on the tank floor
- Holders are available for various applications
- Special solutions are available (e.g. for conical flasks)
Spécification
Reaction Tube Holder | |
Fisherbrand Select, Fisherbrand EASY, Elmasonic P, Elmasonic Select, Elmasonic EASY | |
150/300 |