Learn More
RBMS1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marque: Invitrogen PA528561
338.68 EUR valable jusqu'au 2025-03-29
Utilisez le code promo "24111" pour bénéficier de cette offre.
Alert:
Pour que la promotion s’applique, le client doit acheter trois fois le même produit au prix catalogue sur la même commande pour avoir une remise de 33.33%. Il n’y a pas de limite d’achat pour un multiple de 3 produits. PCODE: 24111
Description
Recommended positive controls: Raji, K562. Predicted reactivity: Mouse (97%), Rat (98%), Xenopus laevis (90%), Chicken (94%), Bovine (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a member of a small family of proteins which bind single stranded DNA/RNA. These proteins are characterized by the presence of two sets of ribonucleoprotein consensus sequence (RNP-CS) that contain conserved motifs, RNP1 and RNP2, originally described in RNA binding proteins, and required for DNA binding. These proteins have been implicated in such diverse functions as DNA replication, gene transcription, cell cycle progression and apoptosis. Several transcript variants, resulting from alternative splicing and encoding different isoforms, have been described. A pseudogene for this locus is found on chromosome 12.
Spécification
RBMS1 | |
Polyclonal | |
Unconjugated | |
RBMS1 | |
2600014B10Rik; AI255215; C2orf12; cervical cancer oncogene 4; c-myc gene single strand binding protein 2; EMBL:AAH87094.1}; HCC-4; hypothetical protein LOC526135; MSSP; MSSP1; MSSP-1; MSSP-2; MSSP-3; Rbms1; rbms1 {ECO:0000312; RNA binding motif single stranded interacting protein 1; RNA binding motif, single stranded interacting protein 1; RNA-binding motif, single-stranded-interacting protein 1; SCR2; Single-stranded DNA-binding protein MSSP-1; suppressor of cdc 2 (cdc13) with RNA binding motif 2; suppressor of CDC2 with RNA-binding motif 2; YC1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
5937 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
P29558 | |
RBMS1 | |
Recombinant protein fragment corresponding to a region within amino acids 1 and 249 of Human RBMS1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.