missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RARRES3 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 12065-1-AP-150UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Retinoids exert biologic effects such as potent growth inhibitory and cell differentiation activities and are used in the treatment of hyperproliferative dermatological diseases. These effects are mediated by specific nuclear receptor proteins that are members of the steroid and thyroid hormone receptor superfamily of transcriptional regulators. RARRES1, RARRES2, and RARRES3 are genes whose expression is upregulated by the synthetic retinoid tazarotene. RARRES3 is thought act as a tumor suppressor or growth regulator.Spécification
RARRES3 | |
Polyclonal | |
Unconjugated | |
RARRES3 | |
HRASLS4, PLA1/2 3, RAR responsive protein TIG3, RARRES3, RIG1, TIG3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
5920 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.37 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9UL19 | |
RARRES3 | |
RARRES3 Fusion Protein Ag2702 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |