Learn More
Thermo Scientific™ Capteur sonique universel Ramsey™ Grad-Line
Tous les produits Thermo Scientific ProduitsDescription
This product has been discontinued – effective June 26, 2023. Aftermarket support including remote technical support and some field service will continue to be available as well as spare parts based on material availability.
Please check for more details on the discontinuation and contact our service team here.
At Thermo Fisher Scientific, we provide our valued customers with the products, solutions, and support that enable you to make the world healthier, cleaner, and safer.
To ensure we can continue to offer innovation, quality, value, and acceptable lead times we continuously evaluate our portfolio and take the tough decisions to discontinue products where necessary.
Spécification
Spécification
| Communications | Numérique |
| Connecteurs | 0.25 tour MS, 6 broches |
| Description | Capteur sonique |
| Courant d’entrée | 0,2 Å |
| Plages de fonctionnement (métrique) | 203 - 813 mm |
| Plages de fonctionnement (impériale) | 8 - 32 po |
| Courant en sortie | 2 Å |
| Alimentation électrique | 8 - 30 VDC |
| Plage de températures | -20°C à +80°C (-4°F à +176°F) |