missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabbit Antiserum anti-Human IgG, Whole Molecule Polyclonal Secondary Antibody, MP Biomedicals
Marque: MP Biomedicals 0855088
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Host Species: Rabbit
Species Reactivity: Hu
Immunostaining
Spécification
Human IgG | |
Unconjugated | |
2 mL | |
Human |
Polyclonal | |
Rabbit | |
Secondary | |
Lyophilized |