missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabbit anti-Mouse IgG (H+L) Secondary Antibody, Alkaline Phosphatase, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Marque: Bio-Techne NB7542
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
IgG (H+L) Polyclonal specifically detects IgG (H+L) in Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Spécification
IgG (H+L) | |
Polyclonal | |
Alkaline Phosphatase (AP) | |
Rabbit | |
Affinity Purified | |
RUO | |
Secondary | |
Mouse | |
IgG |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
1.0 mg/mL | |
Western Blot 1:5000- 1:50000, ELISA 1:5000 - 1:50000, Immunohistochemistry 1:100 - 1:1000, Immunocytochemistry/Immunofluorescence 1:100 - 1:1000, Immunohistochemistry-Paraffin 1:100 - 1:1000 | |
Mouse IgG-heavy and light chain | |
1 mL | |
Adaptive Immunity, Immunology | |
By immunoelectrophoresis and ELISA this antibody reacts specifically with mouse IgG and with light chains common to other mouse immunoglobulins. No antibody was detected against nonimmunoglobulin serum proteins. This antibody may cross react with IgG from other species. | |
Store at 4C. Do not freeze. |
For Research Use Only