missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabbit anti-Bovine IgG (H+L) Secondary Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Marque: Novus Biologicals NB766
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
IgG (H+L) Polyclonal specifically detects IgG (H+L) in Bovine samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Spécification
IgG (H+L) | |
Polyclonal | |
HRP | |
Rabbit | |
Affinity Purified | |
RUO | |
Secondary | |
Bovine | |
IgG |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
1.0 mg/mL | |
Western Blot 1:5000 - 1:50000, ELISA 1:10000- 1:100000, Immunohistochemistry 1:200 - 1:5000, Immunocytochemistry/Immunofluorescence 1:200 - 1:5000, Immunohistochemistry-Paraffin | |
Bovine IgG-heavy and light chain | |
1 mL | |
Adaptive Immunity, Immunology | |
By immunoelectrophoresis and ELISA this reacts specifically with bovine IgG and with light chains common to other bovine immunoglobulins. No was detected against non-immunoglobulin serum proteins. This may cross react with IgG from other species. | |
Store at 4C. Do not freeze. |
For Research Use Only