missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabbit anti-Bovine IgG Fc Secondary Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Marque: Bio-Techne NBP1-73581
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
IgG Fc Polyclonal antibody specifically detects IgG Fc in Bovine samples. It is validated for Western Blot, ELISA, Immunohistochemistry.Spécification
IgG Fc | |
Polyclonal | |
HRP | |
Rabbit | |
Affinity Purified | |
RUO | |
No reaction was observed against Bovine IgG F(ab). | |
Bovine | |
IgG |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
LYOPH | |
Western Blot 1:1000 - 1:10000, ELISA 1:10000 - 1:50000, Immunohistochemistry 1:500 - 1:2500, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin 1:20-1:50 | |
Rabbit anti-Bovine IgG Fc fragment Secondary Antibody was produced by repeated immunization with bovine IgG F(c) fragment in rabbit. | |
1.5 mg | |
Secondary | |
Reconstitute with 1.0 ml deionized water (or equivalent). | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only