missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabbit anti-Bovine IgG F(c) fragment, HRP, Polyclonal, Secondary Antibody, Abnova™
Marque: Abnova PAB10460.1500ug
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Spécification
Rabbit anti-Bovine IgG F(c) fragment | |
Polyclonal | |
Rabbit anti-bovine IgG recognizes bovine IgG F(c) fragment. This secondary antibody was purified using antigen affinity chromatography. The antibody is conjugated with Peroxidase. | |
Lyophilized from 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% gentamicin sulfate) | |
Bovine IgG F(c) fragment | |
1500 μg | |
Bovine | |
Lyophilized |
Dot Blot, ELISA, Immunohistochemistry, Western Blot | |
HRP | |
The optimal working dilution should be determined by the end user. | |
Rabbit | |
This product was prepared from monospecific antiserum by immunoaffinity chromatography, followed by solid phase adsorption(s) to remove any unwanted reactivities. | |
Secondary | |
Store at 4°C on dry atmosphere. After reconstitution with 1 mL of deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
For Research Use Only